3-Iodopyridine, 99%
3-Iodopyridine, 99%
3-Iodopyridine, 99%
Thermo Scientific Chemicals

3-Iodopyridine, 99%

CAS: 1120-90-7 | C5H4IN | 205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 137440010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1120-90-7
IUPAC Name3-iodopyridine
Molecular FormulaC5H4IN
InChI KeyXDELKSRGBLWMBA-UHFFFAOYSA-N
SMILESIC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only