2-Bromo-3'-nitroacetophenone, 97%
2-Bromo-3'-nitroacetophenone, 97%
2-Bromo-3'-nitroacetophenone, 97%
Thermo Scientific Chemicals

2-Bromo-3'-nitroacetophenone, 97%

CAS: 2227-64-7 | C8H6BrNO3 | 244.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 137470050
Price (USD)
22.20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
22.20
Each
Chemical Identifiers
CAS2227-64-7
IUPAC Name2-bromo-1-(3-nitrophenyl)ethan-1-one
Molecular FormulaC8H6BrNO3
InChI KeyGZHPNIQBPGUSSX-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point90°C to 94°C
GC>=96.0 %
Appearance (Color)Yellow-green to beige to light orange
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only