What material is the exterior of 1300 Series Class II, Type A2 Biological Safety Cabinet (Cat. No. 1376) made of?
The exterior of 1300 Series Class II, Type A2 Biological Safety Cabinet (Cat. No. 1376) made of Stainless Steel (304).
Search
View additional product information for 1300 Series Class II, Type A2 Biological Safety Cabinet Packages, 230V 50/60Hz - FAQs (1376)
1 product FAQs found