ESI prepFAST Automated Inline Dilution Systems
Actual product may vary
ESI prepFAST Automated Inline Dilution Systems
ESI prepFAST Automated Inline Dilution Systems
ESI prepFAST Automated Inline Dilution Systems
ESI prepFAST Automated Inline Dilution Systems
Thermo Scientific™
ESI prepFAST Automated Inline Dilution Systems
Automate sample dilution and perform autocalibration to enhance laboratory productivity with the ESI prepFAST system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelFor Use With (Equipment)
1378920ESI SC-4DX prepFASTICP-MS System
BRE0003998ESI SC-2DX prepFASTICP-OES System
BRE0003999ESI SC-4DX prepFASTICP-OES System
BRE0004000ESI SC-8DX prepFASTICP-OES System
BRE0004001ESI SC-14DX prepFASTICP-OES System
1378910ESI SC-2DX prepFASTICP-MS System
1378930ESI SC-8DX prepFASTICP-MS System
1378940ESI SC-14DX prepFASTICP-MS System
BRE0068462ESI 2DXP prepFAST 3iCAP PRO Series ICP-OES, iCAP MX Series ICP-MS
BRE0068464ESI 4DXP prepFAST 3iCAP PRO Series ICP-OES, iCAP MX Series ICP-MS
Catalog number 1378920
Price (MXN)
-
Model:
ESI SC-4DX prepFAST
For Use With (Equipment):
ICP-MS System
Automate sample dilution and perform autocalibration to enhance laboratory productivity with the Elemental Scientific™ (ESI) prepFAST system. This automated sample preparation system combines the DX autosampler, high-throughput FAST technology and a high-precision syringe module to provide automated inline dilution of standards and samples. It saves valuable operator time with autocalibration of the instrument and provides autodilution of overrange and high-matrix samples, all without requiring operator intervention.
Encounter the ultimate in sample introduction automation with the ESI prepFAST automated inline dilution systems. These systems provide high-precision autodilution (1–400x), syringe-driven internal standard addition and high-speed washout. These powerful systems are controlled by the fully integrated software plugin within the Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) Software.
Compatible with:
  • Thermo Scientific™ iCAP™ 6000/7000/PRO Series ICP-OES Systems
  • Thermo Scientific™ iCAP™ Q/RQ/TQ/MX Series ICP-MS Systems
ESI DX prepFAST systems offer automation of three common tasks in trace elemental laboratories
  • Autocalibration—Inline preparation of ICP and ICP-MS calibration standards to improve laboratory efficiency and linearity of calibrations
  • Autodilution—Automated inline dilution of any sample to improve precision and accuracy, reduce vial and reagent usage and accelerate sample throughput and washout
  • Overrange Dilution—Automated dilution and reanalysis of samples containing analyte concentrations above the calibration range or samples that cause internal standard recoveries to fall outside the acceptable range
ESI DX prepFAST system models
  • 2DX prepFAST Autosampler—2 large racks, 4 microplates
  • 4DX prepFAST Autosampler—4 large racks, 6 microplates
  • 8DX prepFAST Autosampler—8 large racks, supplied with Mobile Station
  • 14DX prepFAST Autosampler—14 large racks, supplied with Mobile Station
Specifications
ModelESI SC-4DX prepFAST
For Use With (Equipment)ICP-MS System
TypeDilution System
Unit SizeEach