Tetraethylenepentamine, technical
Tetraethylenepentamine, technical
Tetraethylenepentamine, technical
Thermo Scientific Chemicals

Tetraethylenepentamine, technical

CAS: 112-57-2 | C8H23N5 | 189.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 138110025
Price (USD)
226.65
Special offer
Online exclusive
Ends: 28-Dec-2026
267.00
Save 40.35
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
226.65
Special offer
Online exclusive
Ends: 28-Dec-2026
267.00
Save 40.35
Each
Chemical Identifiers
CAS112-57-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Titration with HCl>=95.0 %
Water=<0.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only