3,4,5,6-Tetrahydrophthalic anhydride, 98%
3,4,5,6-Tetrahydrophthalic anhydride, 98%
3,4,5,6-Tetrahydrophthalic anhydride, 98%
Thermo Scientific Chemicals

3,4,5,6-Tetrahydrophthalic anhydride, 98%

CAS: 2426-02-0 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 138220050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2426-02-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
Infrared spectrumConforms
Melting point70°C to 74°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only