Tetraphenylcyclopentadienone, 99%
Tetraphenylcyclopentadienone, 99%
Tetraphenylcyclopentadienone, 99%
Thermo Scientific Chemicals

Tetraphenylcyclopentadienone, 99%

CAS: 479-33-4 | C29H20O | 384.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 138541000
Price (USD)
335.65
Online exclusive
373.00
Save 37.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
335.65
Online exclusive
373.00
Save 37.35
Each
Chemical Identifiers
CAS479-33-4
IUPAC Nametetraphenylcyclopenta-2,4-dien-1-one
Molecular FormulaC29H20O
InChI KeyPLGPSDNOLCVGSS-UHFFFAOYSA-N
SMILESO=C1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point217°C to 222°C
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only