Thioanisole, 99%
Thioanisole, 99%
Thioanisole, 99%
Thermo Scientific Chemicals

Thioanisole, 99%

CAS: 100-68-5 | C7H8S | 124.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 138675000
Price (USD)
200.65
Online exclusive
223.00
Save 22.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
200.65
Online exclusive
223.00
Save 22.35
Each
Chemical Identifiers
CAS100-68-5
IUPAC Name(methylsulfanyl)benzene
Molecular FormulaC7H8S
InChI KeyHNKJADCVZUBCPG-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5850 to 1.5880 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only