Product Image
Thermo Scientific™

DRI™ Negative Urine Drugs of Abuse Calibrators

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
13881 x 25 mL
16641 x 10 mL
Catalog number 1388
Price (USD)
-
Quantity:
1 x 25 mL
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Negative Urine Drugs of Abuse Calibrators — ready-to-use assays in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0 ng/mL
DescriptionDRI Negative Urine Drugs of Abuse Calibrators
Packaging1 Bottle
Quantity1 x 25 mL
TypeCalibrator
Detectable AnalytesNegative Calibrator
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C