Thiophene, 99+%, extra pure
Thiophene, 99+%, extra pure
Thiophene, 99+%, extra pure
Thermo Scientific Chemicals

Thiophene, 99+%, extra pure

CAS: 110-02-1 | C4H4S | 84.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138825000
Price (EUR)
68,40
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
68,40
Each
Chemical Identifiers
CAS110-02-1
IUPAC Namethiophene
Molecular FormulaC4H4S
InChI KeyYTPLMLYBLZKORZ-UHFFFAOYSA-N
SMILESS1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 % (K.F.)
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<50 APHA
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only