2-Thiophenecarboxaldehyde, 98%
2-Thiophenecarboxaldehyde, 98%
2-Thiophenecarboxaldehyde, 98%
Thermo Scientific Chemicals

2-Thiophenecarboxaldehyde, 98%

CAS: 98-03-3 | C5H4OS | 112.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138830010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-03-3
IUPAC Namethiophene-2-carbaldehyde
Molecular FormulaC5H4OS
InChI KeyCNUDBTRUORMMPA-UHFFFAOYSA-N
SMILESO=CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.5890 to 1.5920 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only