Thiophenol, 99%
Thiophenol, 99%
Thiophenol, 99%
Thermo Scientific Chemicals

Thiophenol, 99%

CAS: 108-98-5 | C6H6S | 110.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Glass Bottle
Catalog number 138870010
Price (USD)
165.65
Special offer
Online exclusive
Ends: 28-Dec-2026
195.00
Save 29.35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
165.65
Special offer
Online exclusive
Ends: 28-Dec-2026
195.00
Save 29.35
Each
Chemical Identifiers
CAS108-98-5
IUPAC Namebenzenethiol
Molecular FormulaC6H6S
InChI KeyRMVRSNDYEFQCLF-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5880 to 1.5910 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only