p-Tolualdehyde, 97%
p-Tolualdehyde, 97%
p-Tolualdehyde, 97%
Thermo Scientific Chemicals

p-Tolualdehyde, 97%

CAS: 104-87-0 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 139001000
Price (USD)
41.65
Online exclusive
45.70
Save 4.05
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online exclusive
45.70
Save 4.05
Each
Chemical Identifiers
CAS104-87-0
IUPAC Name4-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyFXLOVSHXALFLKQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5440 to 1.5480 (20°C, 589 nm)
GC>=96.0 %
Specific gravity(20°C) 1.010 to 1.025
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only