p-Toluenesulfonyl chloride, 99+%
p-Toluenesulfonyl chloride, 99+%
p-Toluenesulfonyl chloride, 99+%
Thermo Scientific Chemicals

p-Toluenesulfonyl chloride, 99+%

CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 139030100
Price (USD)
496.65
Online exclusive
552.00
Save 55.35
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
496.65
Online exclusive
552.00
Save 55.35
Each
Chemical Identifiers
CAS98-59-9
IUPAC Name4-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyYYROPELSRYBVMQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
Melting point67°C to 69°C
Assay>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only