n-Tridecanoic acid, 97%
n-Tridecanoic acid, 97%
n-Tridecanoic acid, 97%
Thermo Scientific Chemicals

n-Tridecanoic acid, 97%

CAS: 638-53-9 | C13H26O2 | 214.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
Packaging:
Glass bottle
Catalog number 139520500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-53-9
IUPAC Nametridecanoic acid
Molecular FormulaC13H26O2
InChI KeySZHOJFHSIKHZHA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or flakes and/or chunks
Infrared spectrumConforms
Melting point40°C to 45°C
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only