Triethyl orthoacetate, 97%
Triethyl orthoacetate, 97%
Triethyl orthoacetate, 97%
Thermo Scientific Chemicals

Triethyl orthoacetate, 97%

CAS: 78-39-7 | C8H18O3 | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 139650025
Price (EUR)
448,00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (EUR)
448,00
Each
Chemical Identifiers
CAS78-39-7
IUPAC Name1,1,1-triethoxyethane
Molecular FormulaC8H18O3
InChI KeyNDQXKKFRNOPRDW-UHFFFAOYSA-N
SMILESCCOC(C)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3950 to 1.3980 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only