Triethyl phosphate, 99%
Triethyl phosphate, 99%
Triethyl phosphate, 99%
Thermo Scientific Chemicals

Triethyl phosphate, 99%

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 139680010
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS78-40-0
IUPAC Nametriethyl phosphate
Molecular FormulaC6H15O4P
InChI KeyDQWPFSLDHJDLRL-UHFFFAOYSA-N
SMILESCCOP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4050 to 1.4070 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only