3,4,5-Trimethoxybenzaldehyde, 99%
3,4,5-Trimethoxybenzaldehyde, 99%
3,4,5-Trimethoxybenzaldehyde, 99%
Thermo Scientific Chemicals

3,4,5-Trimethoxybenzaldehyde, 99%

CAS: 86-81-7 | C10H12O4 | 196.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 139910250
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.70
Save 6.05
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.70
Save 6.05
Each
Chemical Identifiers
CAS86-81-7
IUPAC Name3,4,5-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyOPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to cream
Appearance (Form)Powder or flakes
Melting point73°C to 76°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only