3,4,5-Trimethoxyphenylacetic acid, 99%
3,4,5-Trimethoxyphenylacetic acid, 99%
3,4,5-Trimethoxyphenylacetic acid, 99%
Thermo Scientific Chemicals

3,4,5-Trimethoxyphenylacetic acid, 99%

CAS: 951-82-6 | C11H14O5 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 139990500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS951-82-6
IUPAC Name2-(3,4,5-trimethoxyphenyl)acetic acid
Molecular FormulaC11H14O5
InChI KeyDDSJXCGGOXKGSJ-UHFFFAOYSA-N
SMILESCOC1=CC(CC(O)=O)=CC(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point117°C to 120°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only