CD8a Monoclonal Antibody (4SM15), eBioscience™ - FAQs
CD8a Monoclonal Antibody (4SM15), eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (4SM15), eBioscience™ - FAQs (14-0808-37, 14-0808-80, 14-0808-95, 14-0808-82)
1 product FAQs found
What is the concentration of CD8a Monoclonal Antibody (Cat. No. 14-0808-82)?
The concentration of CD8a Monoclonal Antibody is 0.5 mg/mL.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.