DRI™ THC (Cannabinoid) Drugs of Abuse Calibrators
Product Image
Thermo Scientific™

DRI™ THC (Cannabinoid) Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of DRI THC(Cannabinoid) Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
14001 x 25 mL200 ng/mL
00421 x 5 mL50 ng/mL
00441 x 5 mL100 ng/mL
02061 x 5 mL200 ng/mL
02351 x 5 mL20 ng/mL
13991 x 25 mL100 ng/mL
1398TS1
also known as 1398
1 x 25 mL50 ng/mL
1397TS
also known as 1397
1 x 25 mL20 ng/mL
Catalog number 1400
Price (USD)
-
Request A Quote
Quantity:
1 x 25 mL
Concentration:
200 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ THC(Cannabinoid) Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration200 ng/mL
DescriptionDRI; 200ng/mL; 25mL
Packaging1 Bottle
Quantity1 x 25 mL
TypeCalibrator
Detectable Analytes11-nor-9-COOH-D9-THC
Sample TypeUrine
Unit SizeEach