1,2,4-Trimethylbenzene, 98%
1,2,4-Trimethylbenzene, 98%
1,2,4-Trimethylbenzene, 98%
Thermo Scientific Chemicals

1,2,4-Trimethylbenzene, 98%

CAS: 95-63-6 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 140090025
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 28-Dec-2026
140.00
Save 20.35
Each
Chemical Identifiers
CAS95-63-6
IUPAC Name1,2,4-trimethylbenzene
Molecular FormulaC9H12
InChI KeyGWHJZXXIDMPWGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=97.5 %
Refractive index1.5030 to 1.5050 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only