2,4,4-Trimethyl-1-pentene, 99%
2,4,4-Trimethyl-1-pentene, 99%
2,4,4-Trimethyl-1-pentene, 99%
Thermo Scientific Chemicals

2,4,4-Trimethyl-1-pentene, 99%

CAS: 107-39-1 | C8H16 | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 140185000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-39-1
IUPAC Name2,4,4-trimethylpent-1-ene
Molecular FormulaC8H16
InChI KeyFXNDIJDIPNCZQJ-UHFFFAOYSA-N
SMILESCC(=C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4070 to 1.4090 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only