DL-Tropic acid, 97%
DL-Tropic acid, 97%
DL-Tropic acid, 97%
Thermo Scientific Chemicals

DL-Tropic acid, 97%

CAS: 552-63-6 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 140551000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS552-63-6
IUPAC Name3-hydroxy-2-phenylpropanoic acid
Molecular FormulaC9H10O3
InChI KeyJACRWUWPXAESPB-UHFFFAOYNA-N
SMILESOCC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point115°C to 120°C
Titration with NaOH>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only