Nunc™ Multidishes with Nunclon™ Supra Surface
Thermo Scientific™
Nunc™ Multidishes with Nunclon™ Supra Surface
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Wells
14068112
1406806
14068224
14068348
Catalog number 140681
Price (USD)
496.00
Case of 75
No. of Wells:
12
Price (USD)
496.00
Case of 75
Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc Multidishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
For Laboratory Use
Specifications
DisposableSingle-use
LidWith Lid
No. per Case75 Dishes
No. per Pack1 Dish
ColorClear
MaterialPolystyrene
No. of Wells12
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Supra
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 75