delta-Valerolactam, 99%
delta-Valerolactam, 99%
delta-Valerolactam, 99%
Thermo Scientific Chemicals

delta-Valerolactam, 99%

CAS: 675-20-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 140780050
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Chemical Identifiers
CAS675-20-7
IUPAC Namepiperidin-2-one
Molecular FormulaC5H9NO
InChI KeyXUWHAWMETYGRKB-UHFFFAOYSA-N
SMILESO=C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Low melting crystalline mass
Infrared spectrumConforms
Appearance (Color)White to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only