Valerophenone, 98%
Valerophenone, 98%
Valerophenone, 98%
Thermo Scientific Chemicals

Valerophenone, 98%

CAS: 1009-14-9 | C11H14O | 162.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 140801000
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
141.00
Save 21.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
141.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS1009-14-9
IUPAC Name1-phenylpentan-1-one
Molecular FormulaC11H14O
InChI KeyXKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILESCCCCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5130 to 1.5170 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only