EBSS, no calcium, no magnesium, no phenol red - FAQs
EBSS, no calcium, no magnesium, no phenol red - FAQs
View additional product information for EBSS, no calcium, no magnesium, no phenol red - FAQs (14155048)
1 product FAQs found
Do you offer media without phenol red (phenol red-free media)?
Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.