HBSS, no calcium, no magnesium, 500 mL - FAQs
HBSS, no calcium, no magnesium, 500 mL - FAQs
View additional product information for HBSS, no calcium, no magnesium - FAQs (14170120, 14170112, 14170161)
1 product FAQs found
Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?
The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.