DPBS, no calcium, no magnesium, 500 mL bottle - FAQs
DPBS, no calcium, no magnesium, 500 mL bottle - FAQs
View additional product information for DPBS, no calcium, no magnesium - FAQs (14190094, 14190359, 14190144, 14190342, 14190136, 14190235, 14190367, 14190250)
2 product FAQs found
What is the shelf life of DPBS (Cat. No. 14190136)?
DPBS has a shelf life of 36 months from the date of manufacture, when stored as recommended (15-30 degrees C).
What concentration of EDTA is recommended for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?
We recommend 0.5 mM EDTA prepared in Dulbecco's Phosphate-Buffered Saline (DPBS) without calcium or magnesium (Cat. No. 14190-144; (in Europe, Cat. No. 14190-094)).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.