DPBS (10X), no calcium, no magnesium, 20 x 100 mL - FAQs
View additional product information for DPBS (10X), no calcium, no magnesium - FAQs (14200091)
Error:This product does not have any FAQs available.