ImmunoCAP™ Fruit and Vegetable Allergens, Melon - FAQs
ImmunoCAP™ Fruit and Vegetable Allergens, Melon - FAQs
View additional product information for ImmunoCAP™ Fruit and Vegetable Allergens - FAQs (14481001, 14516110, 14433401, 14518810, 14433601, 14483901, 14429501, 14526910, 14412901, 14445501, 14481201, 14443201, 14481401, 14504610, 14521610, 14484001, 14482901, 14527910, 14421001, 14505110, 14429801, 14421601, 14421801, 14509110, 14522110, 14524410, 14418201, 14528010, 14433101, 14516010, 14433701, 14524510, 14445801, 14443301, 14429601, 14443101, 14481101, 14481301, 14422001, 14506810, 14508910, 14506010, 14484101, 14434601, 14521510, 14504710, 14505210, 14507710, 14421101, 14480901, 14505410, 14421901)
1 product FAQs found
Do you offer a kit for antibody purification?
For purification of antibodies, we offer Melon Gel, which binds other proteins from serum, ascites or culture supernatant, allowing the IgG to be collected in the flow-through. As low pH is typically used to elute IgG from Protein A, G, A/G or L, the use of Melon Gel can spare the IgG from exposure to such harsh conditions, which can be particularly important for monoclonal antibodies.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.