ImmunoCAP™ Epidermal and Animal Protein Allergens
Thermo Scientific™

ImmunoCAP™ Epidermal and Animal Protein Allergens

Measure specific IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon NameRegulatory Status
1443260110 Testse87Rat epithelium, serum proteins and urine proteinsFDA Cleared
1441100116 Testse5Dog danderFDA Cleared
1441090116 Testse1Cat danderFDA Cleared
1441220116 Testse3Horse danderFDA Cleared
1441660116 Testse4Cow danderFDA Cleared
1441680116 Testse7Pigeon droppingsFDA Cleared
1441670116 Testse6Guinea pig epitheliumFDA Cleared
1441690116 Testse70Goose feathersFDA Cleared
1441700110 Testse85Chicken feathersFDA Cleared
1441710110 Testse86Duck feathersFDA Cleared
1443040110 Testse78Budgerigar feathersFDA Cleared
1443080110 Testse82Rabbit epitheliumFDA Cleared
1443030110 Testse77Budgerigar droppingsFDA Cleared
1443070110 Testse81Sheep epitheliumFDA Cleared
1443090110 Testse83Swine epitheliumFDA Cleared
1443060110 Testse80Goat epitheliumFDA Cleared
1443100110 Testse84Hamster epitheliumFDA Cleared
1443270110 Testse88Mouse epithelium, serum proteins and urine proteinsFDA Cleared
1443560110 Testse89Turkey feathersFDA Cleared
1443940110 Testse71Mouse epitheliumFDA Cleared
1443950110 Testse72Mouse urine proteinsFDA Cleared
1443970110 Testse74Rat urine proteinsFDA Cleared
1443960110 Testse73Rat epitheliumFDA Cleared
1443990110 Testse76Mouse serum proteinsFDA Cleared
1443980110 Testse75Rat serum proteinsFDA Cleared
1448240110 Testse201Canary bird feathersFDA Cleared
1450131010 TestsRe214Finch feathersASR
1450151010 TestsRe209Gerbil epitheliumASR
1450161010 TestsRe205Horse, serum proteinsASR
1450341010 TestsRe211Rabbit, urine proteinsASR
1450321010 TestsRe215Pigeon feathersASR
1450331010 TestsRe206Rabbit, serum proteinsASR
1451561010 TestsRe216Deer epitheliumASR
1451571010 TestsRe217Ferret epitheliumASR
1452301010 TestsRe219Chicken, serum proteinsASR
1452291010 TestsRe218Chicken droppingsASR
Catalog number 14432601
Price (USD)
-
Quantity:
10 Tests
Code:
e87
Common Name:
Rat epithelium, serum proteins and urine proteins
Regulatory Status:
FDA Cleared
Quantify specific IgE antibodies with ImmunoCAP™ Epidermal and Animal Protein Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codee87
Common NameRat epithelium, serum proteins and urine proteins
Quantity10 Tests
Code TypeAnimal Protein
Regulatory StatusFDA Cleared
Unit SizeEach