ImmunoCAP™ Tree Pollen Allergens
Thermo Scientific™

ImmunoCAP™ Tree Pollen Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1443740110 Testst73Australian Pine
1441020116 Testst3Common Silver Birch
1441180116 Testst17Japanese Cedar
1441490116 Testst7Oak
1441470116 Testst4Hazel
1441480116 Testst5American Beech
1441460116 Testst2Grey Alder
1441500116 Testst9Olive
1441510116 Testst12Willow
1441610116 Testst1Box Elder
1442250116 Testst6Mountain Juniper
1442260116 Testst8Elm
1442290116 Testst14Cottonwood
1442270116 Testst10Walnut
1442300116 Testst15White Ash
1442320116 Testst18Eucalyptus, Gum-tree
1442330116 Testst19Acacia
1442310116 Testst16White Pine
1442280116 Testst11Maple Leaf Sycamore, London Plane
1442340116 Testst20Mesquite
1442350116 Testst21Melaleuca, Cajeput-tree
1442360116 Testst22Pecan, Hickory
1442370116 Testst23Italian/Mediterranean/Funeral Cypress
1443250110 Testst70Mulberry
1443730110 Testst72Queen Palm
1448230110 Testst208Linden
1448340110 Testst210Privet
1449421010 TestsRt41White Hickory
1449411010 TestsRt37Bald Cypress
1449431010 TestsRt44Hackberry
1449441010 TestsRt45Cedar Elm
1449451010 TestsRt54Russian Olive
1449461010 TestsRt55Scotch Broom
1449471010 TestsRt56Bayberry
1449481010 TestsRt57Red Cedar
1449491010 TestsRt71Red Mulberry
1450021010 TestsRt205Elder
1450051010 TestsRt201Spruce
1451401010 TestsRt206Chestnut
1451481010 TestsRt209Horn Beam
1451461010 TestsRt207Douglas Fir
1451551010 TestsRt211Sweet Gum
1451671010 TestsRt217Peppertree
1451850110 Testst212Cedar
1452081010 TestsRt213Pine
1452181010 TestsRt214Date
1452521010 TestsRt218Virgina Live Oak
1453090110 Testst222Cypress
Catalog number 14437401
Price (USD)
-
Quantity:
10 Tests
Code:
t73
Common Name:
Australian Pine
Quantify specific IgE antibodies with ImmunoCAP™ Tree Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codet73
Common NameAustralian Pine
Quantity10 Tests
Code TypeTree
Regulatory StatusFDA Cleared
Scientific NameCasuarina equisetifolia
Unit SizeEach