ImmunoCAP™ Drug Allergens
Thermo Scientific™

ImmunoCAP™ Drug Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon NameRegulatory Status
1443911010 Testsc70Insulin porcineASR
1441640116 Testsc1Penicilloyl GFDA Cleared
1441650116 Testsc2Penicilloyl VFDA Cleared
1443470110 Testsc73Insulin humanFDA Cleared
1443921010 Testsc71Insulin bovineASR
1444500116 Testsc5AmpicilloylFDA Cleared
1444510116 Testsc6AmoxicilloylFDA Cleared
1448540110 Testsc74Gelatin bovineFDA Cleared
1451631010 TestsRc207ProtamineASR
1451621010 TestsRc206ACTHASR
1452271010 TestsRc208Tetanus toxoidASR
1452661010 Testsc202Suxamethonium (succinylcholine)ASR
1452591010 TestsRc209ChymopapainASR
1453361010 Testsc7CefaclorASR
Catalog number 14439110
Price (USD)
-
Quantity:
10 Tests
Code:
c70
Common Name:
Insulin porcine
Regulatory Status:
ASR
Quantify specific IgE antibodies with ImmunoCAP™ Drug Allergen Components, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codec70
Common NameInsulin porcine
Quantity10 Tests
Code TypeDrug
Regulatory StatusASR
Unit SizeEach