You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Microplates
▸
Cell Culture Microplates
▸
Nunc™ Non-Treated Multidishes
Thermo Scientific™
Nunc™ Non-Treated Multidishes
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Have Questions?
Change view
Catalog Number
No. of Wells
144530
24
150200
12
150239
6
150787
48
179820
4
5 Options
Catalog number
144530
Price (USD)
425.65
Online exclusive
453.00
Save 27.35
Case of 75
Add to cart
No. of Wells:
24
Price (USD)
425.65
Online exclusive
453.00
Save 27.35
Case of 75
Add to cart
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Flat-bottom round wells
Excellent optical properties
Raised well rims to lower the risk of cross-contamination
Lid with notched corners for good fit and easy orientation
Sterile and non-pyrogenic
Warranty
: 90 days
WARNING:
Cancer –
www.P65Warnings.ca.gov
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Includes
Lid
Length (Metric)
128 mm
No. per Case
75
No. per Pack
1
Product Type
Multi-Well Plate
Volume (Metric) Well
1 mL (Well)
Volume (Metric) Working
1 mL
Width (Metric)
86 mm
No. of Wells
24
Sterility
Sterile
Surface Treatment
Untreated
Well Shape
Flat Bottom (F-well)
Unit Size
Case of 75