ImmunoCAP™ Spice Allergens
Thermo Scientific™

ImmunoCAP™ Spice Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCodeCommon Name
14481601f218Paprika, Sweet pepper
14429701f86Parsley
14432901f89Mustard
14481701f280Black pepper
14504410Rf271Anise
14504910Rf278Bay leaf
14504810Rf269Basil
14505710Rf265Caraway
14505810Rf267Cardamon
14506210Rf279Chilipepper
14506310Rf220Cinnamon
14506410Rf268Clove
14506710Rf281Curry (Santa Maria)
14506910Rf277Dill
14507210Rf270Ginger
14507010Rf219Fennel seed
14507510Rf263Green pepper (unripe seed)
14508310Rf266Mace
14508410Rf274Marjoram
14508710Rf282Nutmeg
14508810Rf283Oregano
14510310Rf272Tarragon
14510610Rf234Vanilla
14510510Rf273Thyme
14521410Rf317Coriander
14524810Rf331Saffron
14524910Rf332Mint
14526710Rf339Allspice
14528110Rf344Sage
Catalog number 14481601
Price (USD)
-
Code:
f218
Common Name:
Paprika, Sweet pepper
Quantify specific IgE antibodies with the ImmunoCAP™ Spice Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codef218
Common NamePaprika, Sweet pepper
Quantity10 Tests
Code TypeSpice
Regulatory StatusFDA Cleared
Scientific NameCapsicum annuum
Unit SizeEach