ImmunoCAP™ Food  Allergen Mixes
Thermo Scientific™

ImmunoCAP™ Food Allergen Mixes

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon NameRegulatory Status
1448530110 Testsfx20f4, f5, f6, f9, Wheat, Rye, Barley, RiceFDA Cleared
1442080116 Testsfx3f4, f7, f8, f10, f11, Wheat, Oat, Maize, Sesame seed, BuckwheatFDA Cleared
1442070116 Testsfx2f3, f24, f37, f40, f41, Fish, Shrimp, Blue mussel, Tuna, SalmonFDA Cleared
1442060116 Testsfx1f13, f17, f18, f20, f36, Peanut, Hazel nut, Brazil nut, Almond, CoconutFDA Cleared
1442750116 Testsfx5f1, f2, f3, f4, f13, f14, Egg white, Milk, Fish, Wheat, Peanut, SoybeanFDA Cleared
1443571010 TestsRfx70f272, f274, f273, f275, Tarragon, Marjoram, Thyme, LovageASR
1443581010 TestsRfx71f265, f266, f267, f268, Caraway, Mace, Cardamom, CloveASR
1443591010 TestsRfx72f269, f219, f270, f271, Basil, Fennel seed, Ginger, AniseASR
1448250110 Testsfx73f26, f27, f83, Pork, Beef, ChickenFDA Cleared
1448271010 TestsRfx74f3, f205, f206, f254, Cod, Herring, Mackerel, PlaiceASR
1448440110 Testsfx7f25, f45, f47, f48, f85, Tomato, Yeast, Garlic, Onion, CeleryFDA Cleared
1448450110 Testsfx8f17, f18, f33, f49, f93, Hazel nut, Brazil nut, Orange, Apple, CacaoFDA Cleared
1448460110 Testsfx9f20, f84, f87, f92, f259, Almond, Kiwi, Melon, Banana, GrapeFDA Cleared
1448470110 Testsfx10f26, f27, f75, f83, f284, Pork, Beef, Egg yolk, Chicken, TurkeyFDA Cleared
1448480110 Testsfx13f12, f15, f31, f35, Pea, White bean, Carrot, PotatoFDA Cleared
1448500110 Testsfx15f33, f49, f92, f95, Orange, Apple, Banana, PeachFDA Cleared
1448490110 Testsfx14f25, f214, f216, f218, Tomato, Spinach, Cabbage, PaprikaFDA Cleared
1448510110 Testsfx16f44, f94, f208, f210, Strawberrry, Pear, Lemon, PineappleFDA Cleared
1448520110 Testsfx18f12, f13, f14, Pea, Peanut, SoybeanFDA Cleared
1448560110 Testsfx24f17, f24, f84, f92, Hazel nut, Shrimp, Kiwi, BananaFDA Cleared
1448550110 Testsfx23f26, f27, f83, f284, Pork, Beef, Chicken meat, Turkey meatFDA Cleared
1448570110 Testsfx25f10, f45, f47, f85, Sesame seed, Yeast, Garlic, CeleryFDA Cleared
1451771010 TestsRfx11f8, f12, f15, f31, f260, Maize, Pea, White bean, Carrot, BroccoliASR
1451791010 TestsRfx17f49, f92, f94, f95, Apple, Banana, Pear, PeachASR
1452061010 TestsRfx21f84, f87, f92, f95, f210, Kiwi, Melon, Banana, Peach, PineappleFDA Cleared
1452121010 TestsRfx22f201, f202, f203, f256, Pecan nut, Cashew nut, Pistachio, WalnutASR
Catalog number 14485301
Price (USD)
-
Quantity:
10 Tests
Code:
fx20
Common Name:
f4, f5, f6, f9, Wheat, Rye, Barley, Rice
Regulatory Status:
FDA Cleared

Identify specific IgE antibodies to broad categories of allergens with ImmunoCAP™ Food Allergen Mixes, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to specific allergens. Qualitative measurement of circulating IgE antibodies to allergen mixes provide an assessment of sensitization to group of related allergens. Reflex testing to the individual whole allergens that make up an allergen mix is generally recommended to identify the allergen specific IgE.

ImmunoCAP Specific IgE Mixes offers the laboratory:
  • Detection limit ≥ 0.35 kUA/L
  • Qualitative results
  • Ability to detect specific IgE to multiple allergens in one test
  • Excellent consistency over time, and between countries, systems, labs and persons

Expected test values
Test results for ImmunoCAP Allergen Mixes are derived from multiple allergens and are reported as qualitative values (positive or negative). A cut-off value of 0.35 kUA/L is recommended for ImmunoCAP multiple allergens. Values between limit of quantification and 0.35 kUA/L may represent very low levels of IgE antibodies. Values ≥0.35 kUA/L indicate specific IgE antibodies to one or more of the allergens coupled to ImmunoCAP multiple allergen. Reinvestigation with appropriate ImmunoCAP single allergen is recommended when there is a need to further identify and obtain a quantitative result for the specific allergen(s). A value below 0.35 kUA/L indicates undetectable or very low levels of allergen specific IgE antibodies towards all of the allergens bound to ImmunoCAP multiple allergen but deviations from results obtained with single ImmunoCAP allergens may occur. The interpretation of results obtained with ImmunoCAP multiple allergen cannot be compared with the results with ImmunoCAP single allergen. The degree of positivity of ImmunoCAP multiple allergen cannot be considered the cumulative degree of positivity of the respective ImmunoCAP single allergen.

Specimen collection and preparation
  • Serum and plasma (EDTA or heparin) samples from venous or capillary blood can be used
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2-8°C up to one week, otherwise store at -20°C
  • Avoid repeated freezing and thawing

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:

  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:

  • Severe symptoms including asthma exacerbations—a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

Specifications
Codefx20
Common Namef4, f5, f6, f9, Wheat, Rye, Barley, Rice
Quantity10 Tests
Code TypeFood
Regulatory StatusFDA Cleared
Unit SizeEach