ImmunoCAP™ Seed, Legume and Nut Component Allergens
Thermo Scientific™

ImmunoCAP™ Seed, Legume and Nut Component Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCodeCommon Name
14496501f424rAra h 3 Peanut
14495401f416rTri a 19 Omega-5 Gliadin, Wheat
14496301f422rAra h 1 Peanut
14496401f423rAra h 2 Peanut
14496801f425rCor a 8 LTP, Hazel nut
14498001f427rAra h 9 LTP, Peanut
14498101f428rCor a 1 PR-10, Hazel nut
14499001f431nGly m 5 beta-conglycinin, Soy
14499101f432nGly m 6 Glycinin
14534301f354rBer e 1 Brazil nut
14534001f353rGly m 4 PR-10, Soy
14534101f352rAra h 8 PR-10, Peanut
Catalog number 14496501
Price (USD)
-
Code:
f424
Common Name:
rAra h 3 Peanut

Quantify specific IgE antibodies with ImmunoCAP™ Seed, Legume and Nut Allergen Components down to the molecular level, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen at the molecular level. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show, in general, a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:

  • Truly quantitative measuring range from 0.1-100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons measuring range from 0.1-100 kUA/L

Expected test values
ImmunoCAP Specific IgE Allergen Component results are reported as quantitative values in the range 0.1 to 100 kUA/L, where A represents allergen-specific antibodies. ImmunoCAP Allergen Components are purified proteins, either natural or recombinant. ImmunoCAP Allergen Component tests for specific IgE antibodies employ these purified, individual allergen proteins, to identify and quantify the levels of IgE antibodies at the molecular level. These assays offer the unique opportunity to assess a person's allergic sensitization pattern at the protein level. IgE antibodies to different protein families often elicit different types of reactions, so testing to identify which proteins are inducing the IgE response may indicate if a patient's prior symptoms, or positive test results, were caused by allergy to a given substance or by a cross-reaction with another allergen. Component testing also helps the clinician weigh a patient's risk of a systemic reaction versus a more mild or localized response.

Specimen collection and preparation

  • Serum and plasma (EDTA or heparin) samples from venous or capillary blood can be used
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2-8°C up to one week, otherwise store at -20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:

  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:

  • The allergy march—a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations—a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

Research Use Only. Authorization required.
Specifications
Codef424
Common NamerAra h 3 Peanut
Product TypeSeed, Legume, and Nut Allergen Component
Quantity10 Tests
Code TypeSeed, Legume, and Nut
Regulatory StatusFDA Cleared
Scientific NameArachis hypogaea
Unit SizeEach