ImmunoCAP™ Fish, Shellfish and Mollusk Allergens
Thermo Scientific™

ImmunoCAP™ Fish, Shellfish and Mollusk Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1451591010 TestsRf304Langust (spiny lobster)
1441590116 Testsf3Fish (cod)
1441800116 Testsf23Crab
1441810116 Testsf24Shrimp
1442130116 Testsf37Blue mussel
1442140116 Testsf40Tuna
1442150116 Testsf41Salmon
1442920110 Testsf80Lobster
1444340116 Testsf254Plaice
1444540116 Testsf50Chub mackerel
1444620116 Testsf58Pacific squid
1444630116 Testsf59Octopus
1444650116 Testsf61Sardine, Japanese Pilchard
1444640116 Testsf60Jack mackerel, Scad
1448190110 Testsf204Trout
1448200110 Testsf207Clam
1448370110 Testsf205Herring
1448420110 Testsf290Oyster
1448950110 Testsf338Scallop
1449311010 TestsRf381Red snapper
1449281010 TestsRf147Gulf flounder
1449321010 TestsRf384Whitefish (Inconnu)
1449331010 TestsRf410Grouper
1449301010 TestsRf369Cat fish
1449351010 TestsRf412Orange roughy
1449371010 TestsRf414Tilapia
1449391010 TestsRf42Haddock
1449381010 TestsRf415Walleye pike
1449361010 TestsRf413Pollock
1450191010 TestsRf206Mackerel
1451421010 TestsRf258Squid
1451581010 TestsRf303Halibut
1451941010 TestsRf307Hake
1451951010 TestsRf308Sardine (Pilchard)
1452041010 TestsRf311Megrim
1452051010 TestsRf312Swordfish
1452101010 TestsRf314Snail
1452091010 TestsRf313Anchovy
1452191010 TestsRf320Crayfish
1452561010 TestsRf337Sole
1452831010 TestsRf346Abalone
Catalog number 14515910
Price (USD)
-
Quantity:
10 Tests
Code:
Rf304
Common Name:
Langust (spiny lobster)
Quantify specific IgE antibodies with ImmunoCAP™ Fish, Shellfish and Mollusk Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
CodeRf304
Common NameLangust (spiny lobster)
Quantity10 Tests
Code TypeFish, Shellfish, and Mollusk
Regulatory StatusASR
Scientific NamePalinurus spp.
Unit SizeEach