ImmunoCAP™ Fruit and Vegetable Allergens
Thermo Scientific™

ImmunoCAP™ Fruit and Vegetable Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1451611010 TestsRf301Persimmon
1441290116 Testsf33Orange
1441820116 Testsf25Tomato
1442100116 Testsf31Carrot
1442110116 Testsf35Potato
1442160116 Testsf44Strawberry
1442190116 Testsf48Onion
1442180116 Testsf47Garlic
1442200116 Testsf49Apple
1442960110 Testsf85Celery
1442950110 Testsf84Kiwi
1442980110 Testsf87Melon
1443340110 Testsf92Banana
1443360110 Testsf94Pear
1443310110 Testsf91Mango
1443370110 Testsf95Peach
1443460110 Testsf96Avocado
1444310116 Testsf209Grapefruit
1444320116 Testsf214Spinach
1444330116 Testsf225Pumpkin
1444550116 Testsf51Bamboo shoot
1444580116 Testsf54Sweet potato
1448090110 Testsf259Grape
1448100110 Testsf210Pineapple
1448110110 Testsf242Cherry
1448120110 Testsf237Apricot
1448130110 Testsf215Lettuce
1448140110 Testsf216Cabbage
1448290110 Testsf208Lemon
1448390110 Testsf244Cucumber
1448410110 Testsf260Broccoli
1448400110 Testsf255Plum
1450471010 TestsRf262Aubergine, eggplant
1450461010 TestsRf261Asparagus
1450511010 TestsRf211Blackberry
1450521010 TestsRf288Blueberry
1450601010 TestsRf291Cauliflower
1450541010 TestsRf217Brussel sprouts
1450681010 TestsRf289Date
1450771010 TestsRf292Guava
1450891010 TestsRf293Papaya
1450911010 TestsFf294Passion fruit
1451601010 TestsRf302Mandarin
1451881010 TestsRf306Lime
1452151010 TestsRf318Jack fruit
1452161010 TestsRf319Beetroot
1452211010 TestsRf322Red currant
1452441010 TestsRf328Fig
1452451010 TestsRf329Watermelon
1452691010 TestsRf341Cranberry
1452791010 TestsRf342Olive (black, fresh)
1452801010 TestsRf343Raspberry
Catalog number 14516110
Price (USD)
-
Quantity:
10 Tests
Code:
Rf301
Common Name:
Persimmon
Quantify specific IgE antibodies with ImmunoCAP™ Fruit and Vegetable Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
CodeRf301
Common NamePersimmon
Quantity10 Tests
Code TypeFruit/Vegetable
Regulatory StatusASR
Scientific NameDiospyros kaki
Unit SizeEach