ImmunoCAP™ Weed Pollen Allergens
Thermo Scientific™

ImmunoCAP™ Weed Pollen Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1451921010 TestsRw210Sugar-beet
1441040116 Testsw21Wall pellitory
1441030116 Testsw6Mugwort
1441550116 Testsw8Dandelion
1441530116 Testsw3Giant ragweed
1441540116 Testsw7Marguerite, Ox-eye daisy
1441520116 Testsw2Western ragweed
1441560116 Testsw9Plantain (English), Ribwort
1441570116 Testsw10Goosefoot, Lamb’s quarters
1441620116 Testsw1Common ragweed
1441860116 Testsw19Wall pellitory
1442470116 Testsw4False ragweed
1442480116 Testsw5Wormwood
1442520116 Testsw14Common pigweed
1442490116 Testsw11Saltwort (prickly), Russian thistle
1442500116 Testsw12Goldenrod
1442530116 Testsw15Scale, Lenscale
1442510116 Testsw13Cocklebur
1442550116 Testsw17Firebush (Kochia)
1442540116 Testsw16Rough marshelder
1442560116 Testsw18Sheep sorrel
1442570116 Testsw20Nettle
1449501010 TestsRw23Yellow dock
1449511010 TestsRw45Alfalfa
1449531010 TestsRw82Careless weed
1449521010 TestsRw46Dog fennel
1450241010 TestsRw203Rape
1450221010 TestsRw207Lupin
1450251010 TestsRw204Sunflower
Catalog number 14519210
Price (USD)
-
Quantity:
10 Tests
Code:
Rw210
Common Name:
Sugar-beet
Quantify specific IgE antibodies with ImmunoCAP™ Weed Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
CodeRw210
Common NameSugar-beet
Quantity10 Tests
Code TypeWeed
Regulatory StatusASR
Scientific NameBeta vulgaris
Unit SizeEach