ImmunoCAP™  Seed, Legume and Nut Allergens
Thermo Scientific™

ImmunoCAP™ Seed, Legume and Nut Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1451960110 Testsf309Chick pea
1441150116 Testsf14Soybean
1441130116 Testsf4Wheat
1441260116 Testsf13Peanut
1441250116 Testsf11Buckwheat
1441240116 Testsf8Maize, Corn
1441230116 Testsf5Rye
1441720116 Testsf6Barley
1441730116 Testsf7Oat
1441760116 Testsf12Pea
1441740116 Testsf9Rice
1441750116 Testsf10Sesame seed
1441770116 Testsf15White bean
1441780116 Testsf18Brazil nut
1441790116 Testsf20Almond
1442120116 Testsf36Coconut
1442910110 Testsf79Gluten
1444590116 Testsf55Common millet
1444600116 Testsf56Foxtail millet
1444610116 Testsf57Japanese millet
1445210116 Testsf256Walnut
1445280116 Testsf17Hazel nut
1448070110 Testsf201Pecan nut
1448150110 Testsf235Lentil
1448360110 Testsf203Pistachio
1448350110 Testsf202Cashew nut
1448430110 Testsf299Sweet chestnut
1449291010 TestsRf182Lima bean
1450931010 TestsRf253Pine nut, pignoles
1450971010 TestsRf224Poppy seed
1451001010 TestsRf287Red kidney bean
1451021010 TestsRf227Sugar-beet seed
1450981010 TestsRf226Pumpkin seed
1451871010 TestsRf305Fenugreek
1452111010 TestsRf315Green bean
1452131010 TestsRf316Rape seed
1452501010 TestsRf333Linseed
1452541010 TestsRf335Lupin seed
1452821010 TestsRf345Macadamia nut
1452841010 TestsRf347Quinoa
Catalog number 14519601
Price (USD)
-
Quantity:
10 Tests
Code:
f309
Common Name:
Chick pea
Quantify specific IgE antibodies with ImmunoCAP™ Seed, Legume and Nut Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codef309
Common NameChick pea
Quantity10 Tests
Code TypeSeed, Legume, and Nut
Regulatory StatusFDA Cleared
Scientific NameCicer arietinus
Unit SizeEach