Triphenylmethane, 99+%
Triphenylmethane, 99+%
Triphenylmethane, 99+%
Thermo Scientific Chemicals

Triphenylmethane, 99+%

CAS: 519-73-3 | C19H16 | 244.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 146380250
Price (USD)
35.65
Online exclusive
39.60
Save 3.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Online exclusive
39.60
Save 3.95
Each
Chemical Identifiers
CAS519-73-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point90°C to 94°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only