3-Hexanone, 98%
3-Hexanone, 98%
3-Hexanone, 98%
Thermo Scientific Chemicals

3-Hexanone, 98%

CAS: 589-38-8 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 146900100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-38-8
IUPAC Namehexan-3-one
Molecular FormulaC6H12O
InChI KeyPFCHFHIRKBAQGU-UHFFFAOYSA-N
SMILESCCCC(=O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only