N-Methylcyclohexylamine, 98%
N-Methylcyclohexylamine, 98%
N-Methylcyclohexylamine, 98%
Thermo Scientific Chemicals

N-Methylcyclohexylamine, 98%

CAS: 100-60-7 | C7H15N | 113.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 146990010
Price (USD)
294.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
294.00
Each
Chemical Identifiers
CAS100-60-7
IUPAC NameN-methylcyclohexanamine
Molecular FormulaC7H15N
InChI KeyXTUVJUMINZSXGF-UHFFFAOYSA-N
SMILESCNC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4535 to 1.4575 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only