Pentafluorobenzaldehyde, 98%
Pentafluorobenzaldehyde, 98%
Pentafluorobenzaldehyde, 98%
Thermo Scientific Chemicals

Pentafluorobenzaldehyde, 98%

CAS: 653-37-2 | C7HF5O | 196.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 147090100
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.10
Save 14.45
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
98.10
Save 14.45
Each
Chemical Identifiers
CAS653-37-2
IUPAC Name2,3,4,5,6-pentafluorobenzaldehyde
Molecular FormulaC7HF5O
InChI KeyQJXCFMJTJYCLFG-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C=O)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid after melting
Appearance (Color)Clear yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only