2-Chlorobenzoyl chloride, 98%
2-Chlorobenzoyl chloride, 98%
2-Chlorobenzoyl chloride, 98%
Thermo Scientific Chemicals

2-Chlorobenzoyl chloride, 98%

CAS: 609-65-4 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1471400101 LGlass Bottle
Catalog number 147140010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS609-65-4
IUPAC Name2-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyONIKNECPXCLUHT-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Titration RCOCl>=97.5 %
Refractive index1.5710 to 1.5750 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only