Adenine, 99%
Adenine, 99%
Adenine, 99%
Thermo Scientific Chemicals

Adenine, 99%

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147445000
Price (USD)
1,166.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,372.00
Save 205.35
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,166.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,372.00
Save 205.35
Each
Chemical Identifiers
CAS73-24-5
IUPAC Name7H-purin-6-amine
Molecular FormulaC5H5N5
InChI KeyGFFGJBXGBJISGV-UHFFFAOYSA-N
SMILESNC1=C2NC=NC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Sulfated ash=<0.1 %
HPLC>=98.5 %
Loss on drying=<0.5 % (105°C, 5 hrs)
Appearance (Color)White to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only