Bromomaleic anhydride, 97%
Bromomaleic anhydride, 97%
Bromomaleic anhydride, 97%
Thermo Scientific Chemicals

Bromomaleic anhydride, 97%

CAS: 5926-51-2 | C4HBrO3 | 176.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 147450500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5926-51-2
IUPAC Name3-bromo-2,5-dihydrofuran-2,5-dione
Molecular FormulaC4HBrO3
InChI KeyYPRMWCKXOZFJGF-UHFFFAOYSA-N
SMILESBrC1=CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5410 to 1.5430 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only